Assay Details
| Gene Symbol: Gm10653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003965.2 | 290 | 75 | |||
| GenBank mRNA | AK131930.1 | - | 291 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | feminization 1 homolog b (C. elegans) | Fem1b | mKIAA0396 | -33.8kb | Sense | Chr. 9 - 62791824 - 62811648 on NCBI Build GRCm38 |
| Downstream | calmodulin-like 4 | Calml4 | 2010002G05Rik | 6.7kb | Anti-sense | Chr. 9 - 62857481 - 62875921 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131930.1 transcripts.
Set Membership: |
|

Gm10653
Genomic Map