Assay Details
| Gene Symbol: Btnl5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004051.1 | 1 - 2 | 367 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | butyrophilin-like 4 | Btnl4 |
Btn3a3, EG632126, NG11 |
-18.5kb | Sense | Chr. 17 - 34468410 - 34477428 on NCBI Build GRCm38 |
| Downstream | butyrophilin-like 6 | Btnl6 |
EG624681, Gm6519, NG13 |
10.5kb | Sense | Chr. 17 - 34507935 - 34517352 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Btnl5-ps
Genomic Map