Assay Details
| Gene Symbol: Snord87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004410.1 | 1 | 39 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | minichromosome maintenance domain containing 2 | Mcmdc2 | 6030422M02Rik | -1.5kb | Anti-sense | Chr. 1 - 9906159 - 9940954 on NCBI Build GRCm38 |
| Downstream | transcription factor 24 | Tcf24 |
ENSMUSG00000073738, Gm10567, Gm2330 |
17.6kb | Sense | Chr. 1 - 9960163 - 9968436 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord87
Genomic Map