Assay Details
| Gene Symbol: Npm3-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002702.1 | 1 | 51 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exocyst complex component 6B | Exoc6b |
4930569O18Rik, G430127E12Rik, Sec15b, Sec15l2, mKIAA0919 |
-5.9kb | Anti-sense | Chr. 6 - 84618486 - 85070259 on NCBI Build GRCm38 |
| Downstream | predicted gene 5878 | Gm5878 | EG545861 | 34.3kb | Anti-sense | Chr. 6 - 85111416 - 85126094 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Npm3-ps1
Genomic Map