Assay Details
| Gene Symbol: Gm12070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002890.1 | 1 | 251 | 71 | ||
| GenBank mRNA | BC098095.1 | - | 1 | 251 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vaccinia related kinase 2 | Vrk2 |
2810003O05Rik, AI447698 |
-191.1kb | Anti-sense | Chr. 11 - 26471401 - 26594014 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 85A | Ccdc85a | 1.6mb | Anti-sense | Chr. 11 - 28385684 - 28584441 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12070
Genomic Map