Assay Details
| Gene Symbol: Gm10319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003624.2 | 2 | 349 | 99 | ||
| GenBank mRNA | BC061166.1 | - | 2 | 349 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | murinoglobulin 2 | Mug2 | Cpamd8 | -52.3kb | Sense | Chr. 6 - 122006719 - 122085968 on NCBI Build GRCm38 |
| Downstream |
murinoglobulin, pseudogene 1 |
Mug-ps1 | Mug3 | 37.5kb | Sense | Chr. 6 - 122176053 - 122248566 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10319
Genomic Map