Assay Details
| Gene Symbol: Gm15421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004442.1 | 1 | 147 | 72 | ||
| GenBank mRNA | CK331818.1 | - | 1 | 148 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
origin recognition complex, subunit 5 |
Orc5 |
AL033327, MmORC5, Orc5l |
Sense | Chr. 5 - 22486484 - 22550367 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 6030443J06 gene | 6030443J06Rik | 1700030G06Rik | 21.4kb | Anti-sense | Chr. 5 - 22550413 - 22558171 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15421
Genomic Map