Assay Details
| Gene Symbol: Pdxk-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027316.1 | 6 - 7 | 1562 | 83 | ||
| GenBank mRNA | AK138248.1 | - | 6 - 7 | 1562 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal RNA processing 1 homolog B (S. cerevisiae) | Rrp1b |
2600005C20Rik, D030064A17, Kiaa0179, mKIAA0179 |
-15.9kb | Anti-sense | Chr. 17 - 32036162 - 32062865 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 38585 |
Gm38585 | 32.3kb | Anti-sense | Chr. 17 - 32113924 - 32129237 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Pdxk-ps
Genomic Map