Assay Details
| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027351.1 | 2 - 3 | 472 | 78 | ||
| GenBank mRNA | AK038830.1 | - | 2 - 3 | 474 | 78 | |
| AK141582.1 | - | 429 | 78 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nudix (nucleoside diphosphate linked moiety X)-type motif 22 | Nudt22 |
0610006K04Rik, AI851793, AW545547 |
-581.0 | Anti-sense | Chr. 19 - 6993019 - 6996115 on NCBI Build GRCm38 |
| Downstream | fermitin family homolog 3 (Drosophila) | Fermt3 |
C79673, Kindlin3 |
592.0 | Anti-sense | Chr. 19 - 6998958 - 7019469 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Trpt1
Genomic Map