Assay Details
| Gene Symbol: Gm17769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027377.1 | 1 | 2452 | 97 | ||
| GenBank mRNA | AK141975.1 | - | 1 | 2457 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
adenosine deaminase, RNA-specific, B1 |
Adarb1 |
1700057H01Rik, AW124433, AW558573, Adar2, BB220382, D10Bwg0447e, Red1 |
-103.0 | Anti-sense | Chr. 10 - 77290727 - 77418273 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 207, member A |
Fam207a |
1810008A18Rik, AI303072 |
65.7kb | Anti-sense | Chr. 10 - 77486655 - 77537969 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm17769
Genomic Map