Assay Details
| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027385.1 | 1 - 2 | 78 | 90 | ||
| GenBank mRNA | AK163216.1 | - | 1 - 2 | 80 | 90 | |
| CJ083078.1 | - | 1 - 2 | 78 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon alpha 12 | Ifna12 | -25.4kb | Sense | Chr. 4 - 88602580 - 88603376 on NCBI Build GRCm38 | |
| Downstream | interferon alpha 13 | Ifna13 | 9.5kb | Sense | Chr. 4 - 88643641 - 88644459 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163216.1 transcripts.
Set Membership: |
|

C87499
Genomic Map