Assay Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027491.1 | 1 - 2 | 262 | 74 | ||
| GenBank mRNA | AA199406.1 | - | 1 - 2 | 262 | 74 | |
| AF359558.1 | - | 1 - 2 | 201 | 74 | ||
| BX529138.1 | - | 1 - 2 | 271 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 3A | Kif3a |
Kif3, Kifl, Kns3 |
-9.7kb | Anti-sense | Chr. 11 - 53567195 - 53604248 on NCBI Build GRCm38 |
| Downstream | interleukin 13 | Il13 | Il-13 | 14.0kb | Sense | Chr. 11 - 53631323 - 53634702 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Il4
Genomic Map