Assay Details
| Gene Symbol: Klrb1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024263.1 | 2 - 3 | 237 | 89 | ||
| GenBank mRNA | BC107177.1 | - | 2 - 3 | 218 | 89 | |
| BC107178.1 | - | 2 - 3 | 218 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
C-type lectin domain family 2, member f |
Clec2f |
Clec2f-ps, Clr-c, Clrc, Clrz |
-31.1kb | Sense | Chr. 6 - 129008803 - 129021342 on NCBI Build GRCm38 |
| Downstream |
C-type lectin domain family 2, member e |
Clec2e |
Clra, clr-a |
38.0kb | Anti-sense | Chr. 6 - 129091816 - 129100903 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107177.1, BC107178.1 transcripts.
Set Membership: |
|

Klrb1f
Genomic Map