Assay Details
| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026843.1 | 1 - 2 | 76 | 71 | ||
| GenBank mRNA | BC024582.1 | - | 1 - 2 | 76 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase 7 | Map3k7 |
B430101B05, C87327, Tak1 |
-542.0kb | Anti-sense | Chr. 4 - 31962296 - 32023470 on NCBI Build GRCm38 |
| Downstream |
gap junction protein, alpha 10 |
Gja10 |
Cx-57, Cx59, cx57 |
29.2kb | Sense | Chr. 4 - 32600865 - 32602382 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Bach2os
Genomic Map