Assay Details
| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026942.1 | 3 - 4 | 364 | 92 | ||
| GenBank mRNA | AK087735.1 | - | 3 - 4 | 351 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAB11 family interacting protein 2 (class I) | Rab11fip2 |
4930470G04Rik, A830046J09Rik, AW558126, Nrip11 |
-209.0kb | Anti-sense | Chr. 19 - 59760441 - 59943666 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 204, member A |
Fam204a |
2310065H12Rik, 2610015K05Rik, D19Ertd737e |
38.3kb | Anti-sense | Chr. 19 - 60198586 - 60226726 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK087735.1 transcripts.
Set Membership: |
|

E330013P04Rik
Genomic Map