Assay Details
| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026942.1 | 4 - 5 | 540 | 96 | ||
| GenBank mRNA | AK087735.1 | - | 4 - 5 | 527 | 96 | |
| BC076607.1 | - | 4 - 5 | 540 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAB11 family interacting protein 2 (class I) | Rab11fip2 |
4930470G04Rik, A830046J09Rik, AW558126, Nrip11 |
-216.4kb | Anti-sense | Chr. 19 - 59760441 - 59943666 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 204, member A |
Fam204a |
2310065H12Rik, 2610015K05Rik, D19Ertd737e |
36.0kb | Anti-sense | Chr. 19 - 60198586 - 60226726 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087735.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | AK087735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | BC076607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | NR_026942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | AK087735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | BC076607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497240 | NR_026942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2385083 | AK163106.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167326 | - | - |
| Whole Mouse Genome G4122A | A_51_P497240 | AK087735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497240 | BC076607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497240 | NR_026942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497240 | AK087735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497240 | BC076607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497240 | NR_026942.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14300333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14217399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14217407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14217395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14217401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14300335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14304157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14495428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14321408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14231580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14298302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14501868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14371607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14370777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14314408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14440677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14246968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14485055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14169256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32223699 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10464404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464402 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440000_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10464400 | - | - |
| Mouse Genome 430 2.0 Array | 1440000_at | - | - |
| Mouse Genome 430 2.0 Array | 1440000_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900007420.mm.1 | NR_026942.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007420.mm.1 | NR_026942.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007420.mm.1 | TC1900000859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900007419.mm.1 | NR_026942.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007419.mm.1 | NR_026942.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007419.mm.1 | TC1900000859.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440000_at | - | - |
| Mouse Expression Set 430 | 1440000_PM_at | - | - |
| Murine Genome U74v2 Set | 130408_at | - | - |
| Murine Genome U74v2 Set | 130408_PM_at | - | - |
| Murine Genome U74v2 Set | 169343_PM_at | - | - |
| Murine Genome U74v2 Set | 169343_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440000_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440000_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 402546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100035 | - | - |

E330013P04Rik
Genomic Map