Assay Details
| Gene Symbol: Gm3219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027380.1 | 1 | 153 | 97 | ||
| GenBank mRNA | AW543308.2 | - | 1 | 97 | ||
| CK329453.1 | - | 1 | 97 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate dehydrogenase 1 | Glud1 |
AI118167, Gdh-X, Glud, Gludl |
-22.0 | Sense | Chr. 14 - 34310727 - 34345033 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 25, member C |
Fam25c |
2200001I15Rik, Fam25 |
6.2kb | Anti-sense | Chr. 14 - 34351882 - 34355393 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm3219
Genomic Map