Assay Details
| Gene Symbol: AU022252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012400.2 | NP_001012400.1 | 2 - 3 | 258 | 85 | |
| GenBank mRNA | AK158349.1 | - | 2 - 3 | 216 | 85 | |
| AK167445.1 | - | 2 - 3 | 220 | 85 | ||
| BC035522.1 | - | 2 - 3 | 241 | 85 | ||
| BC089614.1 | - | 2 - 3 | 240 | 6748514 | 85 | |
More Information
Set Membership: |
|

AU022252
Genomic Map