Assay Details
| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145096.1 | NP_001138568.1 | 11 - 12 | 806 | 92 | |
| GenBank mRNA | AK158329.1 | - | 11 - 12 | 808 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hhla1
Genomic Map