Assay Details
| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145096.1 | NP_001138568.1 | 13 - 14 | 1269 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhla1| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145096.1 | NP_001138568.1 | 13 - 14 | 1269 | 64 | |
Set Membership: |
|