Assay Details
| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145096.1 | NP_001138568.1 | 16 - 17 | 1570 | 97 | |
| GenBank mRNA | AK158329.1 | - | 16 - 17 | 1572 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983393 | NM_001145096.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983393 | AK158329.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494882 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104482 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983393 | NM_001145096.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983393 | AK158329.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104482 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08398027 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08397988 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08397970 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08398035 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08398008 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08398008 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08397970 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12029283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12029286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12350700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12329027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12326461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12029292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12364685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11915308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12364680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11855438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12410189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12029278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11999438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30289011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12029284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12364684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30288891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12232858 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429070 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429064 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500012332.mm.1 | NM_001145096.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012332.mm.1 | NM_001145096.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012332.mm.1 | TC1500001644.mm.1 | MTA TC |

Hhla1
Genomic Map