Assay Details
| Gene Symbol: Atp6ap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145879.1 | NP_001139351.1 | 3 - 4 | 257 | 94 | |
| GenBank mRNA | CJ050414.1 | - | 3 - 4 | 261 | 94 | |
| CX238982.1 | - | 3 - 4 | 198 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6ap1l
Genomic Map