Assay Details
| Gene Symbol: Atp6ap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145879.1 | NP_001139351.1 | 4 - 5 | 474 | 99 | |
| GenBank mRNA | CX238982.1 | - | 4 - 5 | 415 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6ap1l
Genomic Map