Assay Details
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159551.1 | NP_001153023.1 | 2 - 3 | 400 | 92 | |
NM_001159552.1 | NP_001153024.1 | 2 - 3 | 400 | 92 | ||
NM_001159553.1 | NP_001153025.1 | 2 - 3 | 400 | 92 | ||
NM_010376.4 | NP_034506.1 | 2 - 3 | 400 | 92 | ||
GenBank mRNA | AF483214.1 | - | 2 - 3 | 367 | 92 | |
AF515662.1 | - | 2 - 3 | 279 | 92 | ||
AJ345032.1 | - | 2 - 3 | 410 | 92 | ||
AK004690.1 | - | 2 - 3 | 374 | 92 | ||
AK007664.1 | - | 2 - 3 | 382 | 92 | ||
AK012600.1 | - | 2 - 3 | 393 | 92 | ||
AK019877.1 | - | 2 - 3 | 374 | 92 | ||
AK028109.1 | - | 2 - 3 | 369 | 92 | ||
AK039008.1 | - | 2 - 3 | 374 | 92 | ||
AK049101.1 | - | 2 - 3 | 374 | 92 | ||
AK050664.1 | - | 2 - 3 | 343 | 92 | ||
AK051151.1 | - | 2 - 3 | 352 | 92 | ||
AK080966.1 | - | 2 - 3 | 362 | 92 | ||
AK159176.1 | - | 2 - 3 | 374 | 92 | ||
AK169922.1 | - | 2 - 3 | 357 | 92 | ||
AK171790.1 | - | 2 - 3 | 328 | 92 | ||
BC056977.1 | - | 2 - 3 | 399 | 6818912 | 92 | |
CA321999.1 | - | 2 - 3 | 400 | 92 | ||
DQ168449.1 | - | 2 - 3 | 416 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
