Assay Details
Gene Symbol: 1600002D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040484.1 | 1 - 2 | 61 | 96 | ||
GenBank mRNA | AK005393.1 | - | 1 - 2 | 62 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
E26 avian leukemia oncogene 2, 3' domain |
Ets2 |
AU022856, Ets-2 |
-122.2kb | Anti-sense | Chr. 16 - 95702407 - 95721049 on NCBI Build GRCm38 |
Downstream |
proteasome (prosome, macropain) assembly chaperone 1 |
Psmg1 |
AW552102, Dscr2 |
50.9kb | Sense | Chr. 16 - 95979935 - 95990903 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
