Assay Details
| Gene Symbol: 1600002D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040484.1 | 3 - 4 | 222 | 89 | ||
| GenBank mRNA | AK005393.1 | - | 3 - 4 | 223 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
E26 avian leukemia oncogene 2, 3' domain |
Ets2 |
AU022856, Ets-2 |
-120.8kb | Anti-sense | Chr. 16 - 95702407 - 95721049 on NCBI Build GRCm38 |
| Downstream |
proteasome (prosome, macropain) assembly chaperone 1 |
Psmg1 |
AW552102, Dscr2 |
137.7kb | Sense | Chr. 16 - 95979935 - 95990903 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1600002D24Rik
Genomic Map