Assay Details
| Gene Symbol: 1700010K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005839.1 | - | 1 - 2 | 105 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40123 |
Gm40123 | -9.4kb | Anti-sense | Chr. 3 - 108226808 - 108239467 on NCBI Build GRCm38 | |
| Downstream | sortilin 1 | Sort1 |
2900053A11Rik, AI852375, Ntr3, Ntsr3 |
26.0kb | Anti-sense | Chr. 3 - 108283026 - 108361519 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700010K24Rik
Genomic Map