Assay Details
| Gene Symbol: 1700010L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005841.1 | - | 48 | 61 | ||
| AK131796.1 | - | 436 | 61 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 186 | Ccdc186 |
1810028B20Rik, A630007B06Rik, AI159700, Otg1 |
Anti-sense | Chr. 19 - 56787485 - 56822482 on NCBI Build GRCm38 | |
| Downstream | tudor domain containing 1 | Tdrd1 | MTR-1 | 3.6kb | Sense | Chr. 19 - 56826171 - 56870012 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700010L13Rik
Genomic Map