Assay Details
| Gene Symbol: 1700012M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005927.1 | - | 1 | 354 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D630024D03 gene | D630024D03Rik | -45.6kb | Sense | Chr. 11 - 31800688 - 31824547 on NCBI Build GRCm38 | |
| Downstream | cytoplasmic polyadenylation element binding protein 4 | Cpeb4 |
Cpe-bp4, Cpeb-4, mCPEB-4 |
139.0 | Anti-sense | Chr. 11 - 31870884 - 31935635 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700012M20Rik
Genomic Map