Assay Details
| Gene Symbol: 1700012O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI449821.1 | - | 82 | |||
| AK005929.1 | - | 351 | 82 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | U box domain containing 5 | Ubox5 |
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5 |
Anti-sense | Chr. 2 - 130589996 - 130630038 on NCBI Build GRCm38 | |
| Downstream | FAST kinase domains 5 | Fastkd5 |
C78212, mKIAA1792 |
23.8kb | Anti-sense | Chr. 2 - 130613838 - 130630027 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700012O15Rik
Genomic Map