Assay Details
Gene Symbol: 1700013A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005935.1 | - | 1 | 451 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin-like 3 | Ubl3 |
AW108023, HCG |
-429.0 | Sense | Chr. 5 - 148504631 - 148552788 on NCBI Build GRCm38 |
Downstream |
predicted gene, 36186 |
Gm36186 | 157.6kb | Anti-sense | Chr. 5 - 148711466 - 148718397 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
