Assay Details
| Gene Symbol: 1700016H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006032.1 | - | 1 | 349 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102640042 | LOC102640042 | -24.7kb | Sense | Chr. 19 - 45311291 - 45338477 on NCBI Build GRCm38 | |
| Downstream | beta-transducin repeat containing protein | Btrc |
Beta-Trcp1, E3RS-IkappaB, E3RSIkappaB, FWD1, Fbw1a, HOS, Slimb, b-TrCP, beta-TrCP |
65.0 | Anti-sense | Chr. 19 - 45363714 - 45533343 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700016H03Rik
Genomic Map