Assay Details
| Gene Symbol: 1700016J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006037.1 | - | 1 - 2 | 44 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | armadillo repeat containing 2 | Armc2 | 2610018I05Rik | Anti-sense | Chr. 10 - 41907755 - 42018382 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 40630 |
Gm40630 | 22.7kb | Anti-sense | Chr. 10 - 42041555 - 42081937 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700016J18Rik
Genomic Map