Assay Details
| Gene Symbol: 1700016L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006046.1 | - | 1 | 152 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae) | Bub3 |
AU019800, AU021329, AU043350, AW146323, C78067 |
-366.0kb | Sense | Chr. 7 - 131560370 - 131571898 on NCBI Build GRCm38 |
| Downstream | G protein-coupled receptor 26 | Gpr26 |
9630036A11Rik, A730021D22 |
28.0kb | Sense | Chr. 7 - 131966135 - 131985633 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700016L15Rik
Genomic Map