Assay Details
| Gene Symbol: 1700016P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006053.1 | - | 1 | 256 | 89 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CA466069.1 | - | 1 | Chr. 1 - 73022554 - 73025499 [+] on Build GRCm38 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transition protein 1 | Tnp1 |
Stp-1, TP1, Tp-1 |
-6.6kb | Sense | Chr. 1 - 73015074 - 73015899 on NCBI Build GRCm38 |
| Downstream | pregnancy induced noncoding RNA | Pinc | 368.5kb | Sense | Chr. 1 - 73391385 - 73407569 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700016P22Rik
Genomic Map