Assay Details
| Gene Symbol: 4930509G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006093.1 | - | 1 | 47 | 96 | |
| AK015735.1 | - | 1 | 1295 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105246071 | LOC105246071 | Sense | Chr. 16 - 11177392 - 11180565 on NCBI Build GRCm38 | ||
| Downstream | ribosomal L1 domain containing 1 | Rsl1d1 |
2410005K20Rik, C76439, pBK1 |
15.3kb | Anti-sense | Chr. 16 - 11193037 - 11203292 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930509G22Rik
Genomic Map