Assay Details
| Gene Symbol: 1700019L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006130.1 | - | 1 | 1450 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cysteine-serine-rich nuclear protein 1 | Csrnp1 |
4931429D10Rik, Axud1, CSRNP-1, URAX1, taip-3 |
Anti-sense | Chr. 9 - 119971163 - 119985134 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 33800 |
Gm33800 | 615.0 | Sense | Chr. 9 - 119985406 - 120011808 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700019L13Rik
Genomic Map