Assay Details
| Gene Symbol: 1700020B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006142.1 | - | 1 | 243 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4737 | Gm4737 |
AL024110, EG11615, ENSMUSG00000048087 |
-28.1kb | Sense | Chr. 16 - 46153629 - 46155018 on NCBI Build GRCm38 |
| Downstream | poliovirus receptor-related 3 | Pvrl3 |
2610301B19Rik, 3000002N23Rik, 4921513D19Rik, AA407785, AU016832, AW538082, CD113 |
211.3kb | Sense | Chr. 16 - 46394858 - 46498684 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700020B03Rik
Genomic Map