Assay Details
| Gene Symbol: 1700021N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045800.1 | 2 - 3 | 300 | 101 | ||
| GenBank mRNA | AK006224.1 | - | 2 - 3 | 301 | 101 | |
| BY706310.2 | - | 2 - 3 | 304 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | platelet-activating factor acetylhydrolase 2 | Pafah2 |
2310074E22Rik, AI507170 |
-21.4kb | Anti-sense | Chr. 4 - 134396320 - 134427412 on NCBI Build GRCm38 |
| Downstream | stathmin 1 | Stmn1 |
19k, Lag, Lap18, Op18, P18, P19, Pig, Pp17, Pp18, Pp19, Pr22, Smn |
18.5kb | Anti-sense | Chr. 4 - 134468320 - 134473843 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700021N21Rik
Genomic Map