Assay Details
| Gene Symbol: 1700023A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006259.1 | - | 1 | 851 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PR domain containing 10 | Prdm10 |
Gm1112, tristanin |
-3.3kb | Anti-sense | Chr. 9 - 31278780 - 31381731 on NCBI Build GRCm38 |
| Downstream | nuclear factor related to kappa B binding protein | Nfrkb | A530090G11Rik | 115.0 | Anti-sense | Chr. 9 - 31386161 - 31421333 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700023A20Rik
Genomic Map