Assay Details
| Gene Symbol: 1700023G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006276.1 | - | 1 | 453 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | a disintegrin and metallopeptidase domain 12 (meltrin alpha) | Adam12 |
Mltna, mKIAA4001 |
-29.2kb | Sense | Chr. 7 - 133883196 - 134232149 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 7, ERATO Doi 443, expressed |
D7Ertd443e |
4933400E14Rik, FATS, Gm497 |
4.4kb | Sense | Chr. 7 - 134266262 - 134520141 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700023G08Rik
Genomic Map