Assay Details
| Gene Symbol: 1700026J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006390.1 | - | 1 - 2 | 641 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger protein 862, pseudogene |
Zfp862-ps |
4732460K03Rik, AI448302, Zfp862, mKIAA0543 |
-1.8kb | Anti-sense | Chr. 6 - 48504339 - 48534832 on NCBI Build GRCm38 |
| Downstream |
ATPase, H+ transporting, lysosomal V0 subunit E2 |
Atp6v0e2 |
0610006O14Rik, NM9.2 |
95.0 | Anti-sense | Chr. 6 - 48537569 - 48541801 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700026J14Rik
Genomic Map