Assay Details
| Gene Symbol: 1700041L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006675.1 | - | 1 | 445 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol glycan anchor biosynthesis, class V |
Pigv |
B330013B03, D430024F16Rik |
Anti-sense | Chr. 4 - 133659914 - 133673997 on NCBI Build GRCm38 | |
| Downstream | AT rich interactive domain 1A (SWI-like) | Arid1a |
1110030E03Rik, BAF250a, Osa1, Smarcf1 |
5.8kb | Anti-sense | Chr. 4 - 133679008 - 133753981 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700041L08Rik
Genomic Map