Assay Details
| Gene Symbol: Spanxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006717.1 | - | 1 | 47 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat and fibronectin type III domain containing 5 | Lrfn5 |
AI427653, AI604817, C130061B21, mKIAA4208 |
-829.9kb | Sense | Chr. 12 - 61522817 - 61857930 on NCBI Build GRCm38 |
| Downstream | fibrous sheath CABYR binding protein | Fscb | EG623046 | 1.8mb | Anti-sense | Chr. 12 - 64471333 - 64474898 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spanxn4
Genomic Map