Assay Details
| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006751.1 | - | 1 | 151 | 98 | |
| CN833883.1 | - | 98 | ||||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T cell receptor alpha chain | Tcra | Tcralpha | -29.6kb | Sense | Chr. 14 - 52427967 - 54224198 on NCBI Build GRCm38 |
| Downstream | abhydrolase domain containing 4 | Abhd4 |
1110035H23Rik, AI429574, Abh4 |
62.0 | Sense | Chr. 14 - 54254129 - 54269169 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dad1
Genomic Map