Assay Details
| Gene Symbol: 1700054M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045919.1 | 1 - 2 | 23 | 54 | ||
| GenBank mRNA | AK006785.1 | - | 1 - 2 | 23 | 54 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 176 | Gpr176 | Gm1012 | Anti-sense | Chr. 2 - 118277096 - 118373419 on NCBI Build GRCm38 | |
| Downstream | eukaryotic translation initiation factor 2 alpha kinase 4 | Eif2ak4 |
2610011M03, GCN2, MGCN2 |
82.6kb | Sense | Chr. 2 - 118388578 - 118475234 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700054M17Rik
Genomic Map