Assay Details
| Gene Symbol: 1700056I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006813.1 | - | 1 | 455 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 30 (zinc transporter), member 4 |
Slc30a4 |
Znt4, lm, znT-4 |
-877.0 | Sense | Chr. 2 - 122681233 - 122702663 on NCBI Build GRCm38 |
| Downstream |
biogenesis of lysosomal organelles complex-1, subunit 6, pallidin, opposite strand |
Bloc1s6os | 4930580A09Rik | 28.8kb | Sense | Chr. 2 - 122732938 - 122738423 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700056I18Rik
Genomic Map