Assay Details
| Gene Symbol: 1700058M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006833.1 | - | 1 | 50 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | voltage-dependent anion channel 3 | Vdac3 | -15.3kb | Anti-sense | Chr. 8 - 22577075 - 22593813 on NCBI Build GRCm38 | |
| Downstream | dickkopf homolog 4 (Xenopus laevis) | Dkk4 | Dkk-4 | 14.3kb | Sense | Chr. 8 - 22623685 - 22627547 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700058M10Rik
Genomic Map