Assay Details
| Gene Symbol: 1700055K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006845.1 | - | 1 | 452 | 77 | |
| AK018863.1 | - | 1 | 451 | 77 | ||
| AK018893.1 | - | 1 | 452 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transcription factor AP-2, gamma |
Tfap2c |
AA409384, AP2gamma, Ap-2.2, Stra2, Tcfap2c |
-303.9kb | Sense | Chr. 2 - 172545678 - 172558622 on NCBI Build GRCm38 |
| Downstream | bone morphogenetic protein 7 | Bmp7 | OP1 | 4.9kb | Anti-sense | Chr. 2 - 172868012 - 172940321 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700055K11Rik
Genomic Map