Assay Details
| Gene Symbol: 1700092E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007040.1 | - | 1 | 652 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930463O16 gene | 4930463O16Rik | -1.2kb | Sense | Chr. 10 - 84488293 - 84497676 on NCBI Build GRCm38 | |
| Downstream | cytoskeleton-associated protein 4 | Ckap4 |
5630400A09Rik, CLIMP-63, P63 |
26.7kb | Anti-sense | Chr. 10 - 84526305 - 84533888 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700092E16Rik
Genomic Map